(R)-2-Phenyl-2-pivalamidoacetic acid 97%

Supplier: Ambeed
Warning

A479952-1G A479952-250MG A479952-5G
77196-766EA 157.59 USD
77196-766 77196-768 77196-770
(R)-2-Phenyl-2-pivalamidoacetic acid 97%
(R)-2-Phenyl-2-pivalamidoacetic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.27 g/mol
MDL Number: MFCD16380977
CAS Number: 40610-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR