3-(2-Bromophenoxy)propanoic acid 97%

Supplier: Ambeed
Warning

A479866-100MG A479866-1G
77196-758EA 58.71 USD
77196-758 77196-760
3-(2-Bromophenoxy)propanoic acid 97%
3-(2-Bromophenoxy)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
MDL Number: MFCD02295726
CAS Number: 165538-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR