4-Amino-2-chloro-5-nitrobenzonitrile 97%

Supplier: Ambeed
Warning

A479803-100MG A479803-250MG A479803-1G
77196-754EA 41.2 USD
77196-754 77196-756 77617-074
4-Amino-2-chloro-5-nitrobenzonitrile 97%
4-Amino-2-chloro-5-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.57 g/mol
MDL Number: MFCD26401904
CAS Number: 172455-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR