7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%

Supplier: Ambeed
Warning

A477879-100MG A477879-250MG
77169-116EA 42.23 USD
77169-116 77169-118
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₀FN·HCl
MW: 187.65 g/mol
MDL Number: MFCD08461074
CAS Number: 799274-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR