4,5-Dimethoxyphthalonitrile 97%

Supplier: Ambeed
Warning

A477784-100MG A477784-1G A477784-250MG A477784-5G
77196-738EA 9 USD
77196-738 77196-740 77196-742 77196-744
4,5-Dimethoxyphthalonitrile 97%
4,5-Dimethoxyphthalonitrile

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MDL Number: MFCD27931583
CAS Number: 88946-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR