2,2-Dimethylbut-3-ynoic acid 92%

Supplier: Ambeed
Danger

A476622-100MG A476622-1G A476622-250MG A476622-5G
77169-096EA 47.38 USD
77169-096 77169-098 77169-100 77169-102
2,2-Dimethylbut-3-ynoic acid 92%
2,2-Dimethylbut-3-ynoic acid

Precaución: for Research Use Only

Formula: C₆H₈O₂
MW: 112.127 g/mol
MDL Number: MFCD03093337
CAS Number: 56663-76-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR