1-Benzhydrylazetidine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A475835-100G A475835-10G A475835-1G A475835-25G A475835-500G A475835-5G
77169-082EA 87.55 USD
77169-082 77169-084 77169-086 77169-088 77169-090 77169-092
1-Benzhydrylazetidine-3-carbonitrile 97%
1-Benzhydrylazetidine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂
MDL Number: MFCD00158910
CAS Number: 36476-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR