1,2,3,4,5,6,7,8-Octahydroanthracene 95%

Supplier: Ambeed
Danger

A475377-100MG A475377-1G A475377-250MG
77196-720EA 57 USD
77196-720 77196-722 77196-724
1,2,3,4,5,6,7,8-Octahydroanthracene 95%
1,2,3,4,5,6,7,8-Octahydroanthracene

Precaución: for Research Use Only

Formula: C₁₄H₁₈
MW: 186.3
Boiling Pt: 294
Melting Pt: 78
Density: 1.017
MDL Number: MFCD00021091
CAS Number: 1079-71-6
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR