(2-Phenylmethyl-1,3-thiazol-4-yl)methanol 97%

Supplier: Ambeed
Danger

Synonyms: (2-Phenylthiazol-4-yl)methanol, 4-(Hydroxymethyl)-2-phenyl-1,3-thiazole

A474848-100MG A474848-1G A474848-250MG
77169-070EA 15.45 USD
77169-070 77169-072 77169-074
(2-Phenylmethyl-1,3-thiazol-4-yl)methanol 97%
(2-MePhenylthyl-1,3-thiazol-4-yl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₉NOS
MW: 191.2496 g/mol
MDL Number: MFCD02682017
CAS Number: 23780-13-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR