2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride 97%

Supplier: Ambeed
Warning

A474364-1G A474364-250MG
77196-716EA 471 USD
77196-716 77196-718
2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride 97%
2-(Aminomethyl)-3,5-dimethylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂N₂·ClH
MDL Number: MFCD17214191
CAS Number: 1257535-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR