1-Hexyl-3-methylimidazolium iodide 98%

Supplier: Ambeed
Warning

A474172-25G A474172-5G
77196-712EA 135 USD
77196-712 77196-714
1-Hexyl-3-methylimidazolium iodide 98%
1-Hexyl-3-methylimidazolium iodide

Precaución: for Research Use Only

Formula: C₁₀H₁₉IN₂
MDL Number: MFCD12964976
CAS Number: 178631-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR