1-Boc-4-nitroindole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Nitroindole, N-BOC protected, tert-Butyl 4-nitro-1H-indole-1-carboxylate

A472896-1G A472896-250MG
77196-702EA 44.29 USD
77196-702 77196-704
1-Boc-4-nitroindole 95%
1-Boc-4-nitroindole

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₄
MW: 262.27 g/mol
MDL Number: MFCD08689543
CAS Number: 913836-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR