1,3-Diphenylpropane 98%

Supplier: Ambeed
Warning

A471774-1G A471774-25G A471774-5G
77196-686EA 26.78 USD
77196-686 77196-688 77196-690
1,3-Diphenylpropane 98%
1,3-Diphenylpropane

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂CH₂CH₂(C₆H₅)
MW: 196.29 g/mol
MDL Number: MFCD00043574
CAS Number: 1081-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR