7-Bromo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine 97%

Supplier: Ambeed
Warning

A471714-250MG A471714-1G
77169-046EA 39 USD
77169-046 77613-460
7-Bromo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine 97%
7-Bromo-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O
MDL Number: MFCD15527041
CAS Number: 946121-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR