1,3,5-Tris(bromomethyl)-2,4,6-triethylbenzene 95%

Supplier: Ambeed
Danger

A471577-10G A471577-5G A471577-1G
77196-682EA 208 USD
77196-682 77196-684 77568-930
1,3,5-Tris(bromomethyl)-2,4,6-triethylbenzene 95%
1,3,5-Tris(bromomethyl)-2,4,6-triethylbenzene

Precaución: For research use only.

Formula: C₁₅H₂₁Br₃
MDL Number: MFCD01076347
CAS Number: 181058-08-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR