3-Acetyl-6-chloroimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A469966-10G A469966-1G A469966-250MG
77196-668EA 192.61 USD
77196-668 77196-670 77196-672
3-Acetyl-6-chloroimidazo[1,2-b]pyridazine 97%
3-Acetyl-6-chloroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₈H₆ClN₃O
MW: 195.606 g/mol
MDL Number: MFCD11044766
CAS Number: 90734-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR