2,4-Dichlorophenylacetylene 97%

Supplier: Ambeed
Warning

A469276-1G A469276-250MG A469276-100MG
77169-040EA 125 USD
77169-040 77169-042 77892-949
2,4-Dichlorophenylacetylene 97%
2,4-Dichlorophenylacetylene

Precaución: for Research Use Only

Formula: C₈H₄Cl₂
MDL Number: MFCD11655752
CAS Number: 75717-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR