1-((Tetrahydro-2H-pyran-4-yl)methyl)piperidin-4-amine dihydrochloride 97%

Supplier: Ambeed
Warning

A468691-100MG A468691-250MG
77196-660EA 122 USD
77196-660 77196-662
1-((Tetrahydro-2H-pyran-4-yl)methyl)piperidin-4-amine dihydrochloride 97%
1-((Tetrahydro-2H-pyran-4-yl)methyl)piperidin-4-amine dihydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₂₄Cl₂N₂O
MW: 271.23 g/mol
MDL Number: MFCD18378137
CAS Number: 1286274-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR