3-Bromo-1H-pyrrolo[2,3-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-7-azaindole

A467985-100G A467985-10G A467985-1G A467985-25G A467985-5G
77169-030EA 315.18 USD
77169-030 77169-032 77169-034 77169-036 77169-038
3-Bromo-1H-pyrrolo[2,3-b]pyridine 97%
3-Bromo-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD02179276
CAS Number: 74420-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR