2,2-Diethoxyacetic acid 97%

Supplier: Ambeed
Danger

A467352-1G A467352-250MG
77169-018EA 110 USD
77169-018 77169-020
2,2-Diethoxyacetic acid 97%
2,2-Diethoxyacetic acid

Precaución: for Research Use Only

Formula: C₆H₁₂O₄
MDL Number: MFCD06208355
CAS Number: 20461-86-3
UN: 3265
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR