(4-(Methoxycarbonyl)-2-(trifluoromethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A467115-100MG A467115-1G A467115-250MG
77196-642EA 36.05 USD
77196-642 77196-644 77196-646
(4-(Methoxycarbonyl)-2-(trifluoromethyl)phenyl)boronic acid 97%
(4-(Methoxycarbonyl)-2-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₈BF₃O₄
MW: 247.96 g/mol
MDL Number: MFCD28515374
CAS Number: 2377607-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR