4'-(Trifluoromethyl)biphenyl-2-carboxylic acid 98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Trifluoromethyl)-2'-biphenylcarboxylic acid, 4'-(Trifluoromethyl)-2-biphenylcarboxylic acid, 4'-Trifluoromethyl-2-biphenyl carboxylic acid, 4'-(Trifluoromethyl)-[1,1'-biphenyl]-2-carboxylic acid
A466346-10G
A466346-1G
A466346-250MG
A466346-25G
A466346-5G
A466346-100G
77169-008EA
35.02
USD
77169-008
77169-010
77169-012
77169-014
77169-016
77684-254
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid 98%, Ambeed.Inc
4'-(Trifluoromethyl)biphenyl-2-carboxylic acid
Precaución: For research use only.
Formula:
C₁₄H₉F₃O₂ MW: 266.22 g/mol |
MDL Number:
MFCD00075353 CAS Number: 84392-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...