2-Cyano-4-fluorobenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-formylbenzonitrile

A465290-1G A465290-250MG A465290-5G
77196-620EA 50 USD
77196-620 77196-622 77196-624
2-Cyano-4-fluorobenzaldehyde 98%
2-Cyano-4-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄FNO
MW: 149.1219 g/mol
MDL Number: MFCD08275455
CAS Number: 77532-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR