Methyl 1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A463139-100MG A463139-250MG A463139-50MG
77196-606EA 81.37 USD
77196-606 77196-608 77196-610
Methyl 1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylate 97%
Methyl 1-(2,2,2-trifluoroethyl)-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂O₂
MW: 208.13 g/mol
MDL Number: MFCD18262321
CAS Number: 1245772-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR