4-(4-Chlorophenyl)piperidine 97%

Supplier: Ambeed
Warning

A462158-5G A462158-1G
77168-982EA 254.41 USD
77168-982 77551-404
4-(4-Chlorophenyl)piperidine 97%
4-(4-Chlorophenyl)piperidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClN
MW: 195.69 g/mol
MDL Number: MFCD03077010
CAS Number: 26905-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR