Methyl 2,4-dioxopiperidine-3-carboxylate 96%

Supplier: Ambeed
Warning

A460791-100MG A460791-1G A460791-250MG A460791-50MG
77168-972EA 150.38 USD
77168-972 77168-974 77168-976 77168-978
Methyl 2,4-dioxopiperidine-3-carboxylate 96%
Methyl 2,4-dioxopiperidine-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₄
MW: 171.15 g/mol
MDL Number: MFCD16876376
CAS Number: 74730-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR