N-Methyl-1H-pyrrolo[2,3-b]pyridin-6-amine 97%

Supplier: Ambeed
Warning

A460590-100MG A460590-1G A460590-250MG
77168-966EA 63.86 USD
77168-966 77168-968 77168-970
N-Methyl-1H-pyrrolo[2,3-b]pyridin-6-amine 97%
N-Methyl-1H-pyrrolo[2,3-b]pyridin-6-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.17 g/mol
MDL Number: MFCD09910035
CAS Number: 1018441-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR