1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
Supplier: Ambeed
|
Warning
|
A460339-1G
A460339-250MG
A460339-5G
77168-960EA
44.29
USD
77168-960
77168-962
77168-964
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
1-(2-Fluorobenzyl)-1H-pyrazolo[3,4-b]pyridine-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₄H₉FN₄ MW: 252.25 g/mol |
MDL Number:
MFCD20268231 CAS Number: 256376-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...