(3R)-Morpholine-3-carboxylic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Morpholine-3-carboxylic acid hydrochloride

A459926-1G A459926-250MG A459926-5G
77168-940EA 57.68 USD
77168-940 77168-942 77168-944
(3R)-Morpholine-3-carboxylic acid hydrochloride 98%
(3R)-Morpholine-3-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₉NO₃·ClH
MW: 167.591 g/mol
MDL Number: MFCD06809588
CAS Number: 1187928-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR