Boc-2-amino-6-fluorobenzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-((tert-Butoxycarbonyl)amino)-6-fluorobenzoic acid

A458971-100MG A458971-250MG
77196-576EA 14 USD
77196-576 77196-578
Boc-2-amino-6-fluorobenzoic acid 97%
Boc-2-amino-6-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNO₄
MDL Number: MFCD02682192
CAS Number: 1048919-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR