(1S)-N,N-Dimethyl-1-[(2R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine 98%

Supplier: Ambeed
Warning

A458062-100MG A458062-1G A458062-250MG
77196-570EA 72 USD
77196-570 77196-572 77196-574
(1S)-N,N-Dimethyl-1-[(2R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine 98%
(1S)-N,N-Dimethyl-1-[(2R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine

Precaución: for Research Use Only

CAS Number: 119477-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR