1-(2,4-Dichlorophenyl)ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-1-(2,4-dichlorophenyl)ethane, 2,4-Dichloro-alpha-methylbenzylamine, 1-(2,4-Dichlorophenyl)ethanamine

A458060-100G A458060-1G A458060-25G A458060-5G
77196-562EA 317.24 USD
77196-562 77196-564 77196-566 77196-568
1-(2,4-Dichlorophenyl)ethylamine 98%
1-(2,4-Dichlorophenyl)ethylamine

Precaución: for Research Use Only

Formula: C₈H₉Cl₂N
MW: 190.07 g/mol
MDL Number: MFCD05215236
CAS Number: 89981-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR