4-Chloroisatoic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 7-Chloro-1H-benzo[d][1,3]oxazine-2,4-dione, 7-Chloro-2H-3,1-benzoxazine-2,4(1H)-dione

A455973-1G A455973-25G A455973-5G
77196-532EA 8.24 USD
77196-532 77196-534 77594-428
4-Chloroisatoic anhydride 97%
4-Chloroisatoic anhydride

Precaución: for Research Use Only

Formula: C₈H₄ClNO₃
MW: 197.58 g/mol
MDL Number: MFCD00090431
CAS Number: 40928-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR