1-(4-Chlorophenyl)cyclopentanecarboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid

A455746-1G A455746-25G A455746-5G
77196-518EA 16.48 USD
77196-518 77196-520 77196-522
1-(4-Chlorophenyl)cyclopentanecarboxylic acid 95%
1-(4-Chlorophenyl)cyclopentanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClO₂
MW: 224.6834 g/mol
MDL Number: MFCD00001373
CAS Number: 80789-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR