1,4-Dichloro-9,10-anthraquinone 95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Dichloroanthracene-9,10-dione

A454699-1G A454699-5G
77196-514EA 195.7 USD
77196-514 77196-516
1,4-Dichloro-9,10-anthraquinone 95%
1,4-Dichloro-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₆Cl₂O₂
MW: 277.1022 g/mol
MDL Number: MFCD00192002
CAS Number: 602-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR