1-Bromo-8-chloronaphthalene 95%

Supplier: Ambeed
Warning

A454364-100G A454364-1G A454364-250MG A454364-25G A454364-500G A454364-5G
77168-902EA 259 USD
77168-902 77168-904 77168-906 77168-908 77168-910 77168-912
1-Bromo-8-chloronaphthalene 95%
1-Bromo-8-chloronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrCl
MDL Number: MFCD17012231
CAS Number: 20816-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR