Methyl 5-fluoro-1H-indole-3-carboxylate 97%

Supplier: Ambeed
Warning

A453776-100MG A453776-1G A453776-250MG A453776-5G
77168-894EA 24.72 USD
77168-894 77168-896 77168-898 77168-900
Methyl 5-fluoro-1H-indole-3-carboxylate 97%
Methyl 5-fluoro-1H-indole-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.174 g/mol
MDL Number: MFCD06203952
CAS Number: 310886-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR