Cyclobut-1-enecarboxylic acid 95%

Supplier: Ambeed
Warning

A453458-100MG A453458-250MG
77168-892EA 183.34 USD
77168-892 77684-241
Cyclobut-1-enecarboxylic acid 95%
Cyclobut-1-enecarboxylic acid

Precaución: For research use only.

Formula: C₅H₆O₂
MW: 98.09 g/mol
MDL Number: MFCD13192488
CAS Number: 23519-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR