o-Toluic hydrazide 97%

Supplier: Ambeed
Warning

A453444-100G A453444-5G
77196-504EA 86 USD
77196-504 77196-506
o-Toluic hydrazide 97%
o-Toluic hydrazide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O
MDL Number: MFCD00014756
CAS Number: 7658-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR