2,9-Di(naphthalen-2-yl)-4,7-diphenyl-1,10-phenanthroline 95%

Supplier: Ambeed
Warning

A453389-100MG A453389-1G A453389-250MG
77168-886EA 64 USD
77168-886 77168-888 77168-890
2,9-Di(naphthalen-2-yl)-4,7-diphenyl-1,10-phenanthroline 95%
2,9-Di(naphthalen-2-yl)-4,7-diphenyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₄₄H₂₈N₂
MDL Number: MFCD12022443
CAS Number: 1174006-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR