(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-(hydroxymethyl)phenylboronic acid

A451882-100MG A451882-1G A451882-250MG A451882-5G
77196-494EA 49.44 USD
77196-494 77196-496 77196-498 77196-500
(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid 97%
(3-Chloro-4-(hydroxymethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.40 g/mol
MDL Number: MFCD18384123
CAS Number: 1190875-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR