Dodecane-1,12-diamine, N-BOC protected 96%

Supplier: Ambeed
Warning

A450896-100MG A450896-1G A450896-250MG A450896-5G A450896-25G
77196-480EA 61 USD
77196-480 77196-482 77196-484 77196-486 77746-923
Dodecane-1,12-diamine, N-BOC protected 96%
Dodecane-1,12-diamine, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₇H₃₆N₂O₂
MDL Number: MFCD02094503
CAS Number: 109792-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR