1,4-Bis(triethoxysilyl)benzene 98%

Supplier: Ambeed
Warning

A450163-1G A450163-25G A450163-5G
77196-470EA 17.51 USD
77196-470 77196-472 77196-474
1,4-Bis(triethoxysilyl)benzene 98%
1,4-Bis(triethoxysilyl)benzene

Precaución: for Research Use Only

Formula: C₁₈H₃₄O₆Si₂
MW: 402.63 g/mol
MDL Number: MFCD05664351
CAS Number: 2615-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR