4-(Phenylamino)-1H-1,2,3-triazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A448884-100MG A448884-1G A448884-250MG
77196-462EA 163 USD
77196-462 77196-464 77196-466
4-(Phenylamino)-1H-1,2,3-triazole-5-carboxylic acid 97%
4-(Phenylamino)-1H-1,2,3-triazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₄O₂
MDL Number: MFCD18806979
CAS Number: 72693-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR