1-Isobutyl-1H-pyrazol-4-amine 97%

Supplier: Ambeed
Warning

A448361-100MG A448361-1G A448361-250MG
77196-448EA 51.5 USD
77196-448 77196-450 77196-452
1-Isobutyl-1H-pyrazol-4-amine 97%
1-Isobutyl-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₇H₁₃N₃
MW: 139.19 g/mol
MDL Number: MFCD11128192
CAS Number: 405548-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR