4-Deoxypyridoxine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-(hydroxymethyl)-2,4-dimethylpyridin-3-ol hydrochloride, 5-Hydroxy-4,6-dimethyl-3-pyridinemethanol Hydrochloride, 4-Deoxypyridoxine HCl

A447157-100MG A447157-250MG A447157-5G A447157-1G
77196-438EA 22.66 USD
77196-438 77196-440 77746-918 77746-917
4-Deoxypyridoxine hydrochloride 98%
4-Deoxypyridoxine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 189.64 g/mol
MDL Number: MFCD00051296
CAS Number: 148-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR