Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

A446507-100MG A446507-250MG A446507-1G
77168-850EA 35.02 USD
77168-850 77168-852 77684-226
Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 95%
Ethyl-6-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate

Precaución: For research use only.

Formula: C₉H₈BrN₃O₂
MW: 270.08 g/mol
MDL Number: MFCD10574908
CAS Number: 1027511-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR