N-Methoxy-N-methyl-2-phenylacetamide 95%

Supplier: Ambeed
Warning

A445019-1G A445019-250MG A445019-5G
77196-426EA 27.81 USD
77196-426 77196-428 77196-430
N-Methoxy-N-methyl-2-phenylacetamide 95%
N-Methoxy-N-methyl-2-phenylacetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD14707542
CAS Number: 95092-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR