2-Carbamoyl-6-fluorophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

A444585-1G A444585-250MG A444585-5G
77196-410EA 133 USD
77196-410 77196-412 77196-414
2-Carbamoyl-6-fluorophenylboronic acid pinacol ester 97%
2-Carbamoyl-6-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BFNO₃
MDL Number: MFCD18730388
CAS Number: 2096333-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR