1,4-Bis(2-methyl-1H-imidazol-1-yl)butane 95+%

Supplier: Ambeed
Warning

A443716-100MG A443716-1G A443716-250MG A443716-5G
77168-824EA 51.5 USD
77168-824 77168-826 77168-828 77168-830
1,4-Bis(2-methyl-1H-imidazol-1-yl)butane 95+%
1,4-Bis(2-methyl-1H-imidazol-1-yl)butane

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₄
MW: 218.29 g/mol
MDL Number: MFCD29919148
CAS Number: 52550-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR